Skip to Content
Merck
All Photos(1)

Documents

P101

Sigma-Aldrich

2-Phenylaminoadenosine

>97%, solid

Synonym(s):

CV-1808

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N6O4
CAS Number:
Molecular Weight:
358.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

>97%

form

solid

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 3.6 mg/mL
H2O: insoluble
absolute ethanol: slightly soluble
ethanol: water (1:1): soluble
methanol: soluble (hot)

storage temp.

−20°C

SMILES string

Nc1nc(Nc2ccccc2)nc3n(cnc13)[C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O

InChI

1S/C16H18N6O4/c17-13-10-14(21-16(20-13)19-8-4-2-1-3-5-8)22(7-18-10)15-12(25)11(24)9(6-23)26-15/h1-5,7,9,11-12,15,23-25H,6H2,(H3,17,19,20,21)/t9-,11-,12-,15-/m1/s1

InChI key

SCNILGOVBBRMBK-SDBHATRESA-N

Gene Information

Biochem/physiol Actions

Selective A2 adenosine receptor agonist; potent coronary vasodilator; weak inhibitor of adenosine uptake by rat cerebral cortical synaptosomes.

Reconstitution

Solutions should be freshly prepared.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Phillis et al.
Pharmacology, biochemistry, and behavior, 24(2), 263-270 (1986-02-01)
Mice were implanted with chronic indwelling cannulae in the lateral cerebral ventricle. A series of adenosine analogs and related compounds were injected into the lateral ventricle (ICVT) and their effects on spontaneous locomotor activity recorded. All analogs produced dose-related decreases
D A Taylor et al.
European journal of pharmacology, 85(3-4), 335-338 (1982-12-03)
2-Chloroadenosine (2-CADO) and 2-phenylaminoadenosine (CV 1808) were compared in a CNS purinergic receptor binding assay and the inhibition of neurogenic contractions of the vas deferens. Both 2-CADO and CV 1808 are more potent than adenosine in both preparations. CV 1808
P H Wu et al.
General pharmacology, 15(3), 251-254 (1984-01-01)
Various adenosine derivatives, methylxanthines and other compounds were tested for their abilities to inhibit the rapid uptake of adenosine by rat cerebral cortical synaptosomes. Several pharmacologically potent derivatives of adenosine were weak inhibitors of uptake with IC20 values in excess
Mark Soave et al.
SLAS discovery : advancing life sciences R & D, 25(2), 186-194 (2019-10-05)
Receptor internalization in response to prolonged agonist treatment is an important regulator of G protein-coupled receptor (GPCR) function. The adenosine A1 receptor (A1AR) is one of the adenosine receptor family of GPCRs, and evidence for its agonist-induced internalization is equivocal.
Stimulation of Adenosine A
Sarawuth Phosri et al.
Frontiers in pharmacology, 8, 428-428 (2017-07-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service