Skip to Content
Merck
All Photos(1)

Key Documents

OH30129357

Ohaus® pH Electrode

ST260

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115604

material

epoxy shaft

packaging

pack of 1 ea

manufacturer/tradename

Ohaus® 30129357

pH measuring range

0.00-14 pH unit

temperature measuring range

0.0-100 °C

Looking for similar products? Visit Product Comparison Guide

General description

All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.

Application

pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement

Features and Benefits

Can be used in conjunction with all Starter bench and portable meters

Legal Information

Ohaus is a registered trademark of Ohaus Corp.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service