Skip to Content
Merck
All Photos(2)

Key Documents

O6501

Sigma-Aldrich

Arachidyl oleate

~99%, liquid

Synonym(s):

Oleic acid arachidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H74O2
CAS Number:
Molecular Weight:
562.99
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99%

form

liquid

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCCCOC(=O)CCCCCCC\C=C/CCCCCCCC

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianzhong Chen et al.
Lipids, 50(8), 821-836 (2015-07-17)
A series of different types of wax esters (represented by RCOOR') were systematically studied by using electrospray ionization (ESI) collision-induced dissociation tandem mass spectrometry (MS/MS) along with pseudo MS(3) (in-source dissociation combined with MS/MS) on a quadrupole time-of-flight (Q-TOF) mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service