Skip to Content
Merck
All Photos(1)

Key Documents

O5129

Supelco

Oxazepam-d5

Synonym(s):

7-Chloro-1,3-dihydro-3-hydroxy-5-(phenyl-d5)-2H-1,4-benzodiazepin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H6D5ClN2O2
CAS Number:
Molecular Weight:
291.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

isotopic purity

98 atom % D

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2=NC(O)C(=O)Nc3ccc(Cl)cc23

InChI

1S/C15H11ClN2O2/c16-10-6-7-12-11(8-10)13(9-4-2-1-3-5-9)18-15(20)14(19)17-12/h1-8,15,20H,(H,17,19)/i1D,2D,3D,4D,5D

InChI key

ADIMAYPTOBDMTL-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernhard Watzer et al.
International journal of legal medicine, 129(4), 759-769 (2014-12-20)
Arthrotec(®) (AT) is a combination of diclofenac, a nonsteroidal anti-inflammatory drug (NSAID), and misoprostol (MP), a synthetic analogue of prostaglandin E1 (PGE1). MP is a lipophilic methyl ester prodrug. It is readily metabolized to the biologically active misoprostol acid (MPA).
André Valle de Bairros et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 975, 24-33 (2014-12-03)
In this study, it is shown a method for the determination of benzodiazepines and their main metabolites in urine samples by hollow-fiber liquid-phase microextraction (LPME) in the three-phase mode. Initially, the hydrolysis step was performed using 100 μL of sodium
Niclas Stephanson et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 985, 189-196 (2015-02-18)
A mass spectrometric method for drugs of abuse testing in exhaled breath employing a sampling device collecting aerosol particles was developed and applied in routine use. Analytes covered were amphetamine, methamphetamine, 6-acetylmorphine, morphine, cocaine, benzoylecgonine, diazepam, oxazepam and tetrahydrocannabinol. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service