Skip to Content
Merck
All Photos(1)

Key Documents

O2501

Sigma-Aldrich

Ornithine Transcarbamylase from Streptococcus faecalis

lyophilized powder, ≥300 units/mg protein

Synonym(s):

Carbamoylphosphate:L-Ornithine Carbamoyltransferase, OTCase, Ornithine Carbamoyl Transferase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

Quality Level

specific activity

≥300 units/mg protein

composition

Protein, ≥65% Lowry

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mutations in ornithine transcarbamylase cause Ornithine transcarbamylase deficiency which prevents the clearance of the byproduct ammonia leading to hyperammonemia. [1]

Unit Definition

One unit will form 1.0 μmole of citrulline from ornithine and carbamyl phosphate per min at pH 8.5 at 37 °C.

Physical form

Lyophilized powder contains Tris buffer salts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatjana Skrbić et al.
PLoS computational biology, 8(6), e1002504-e1002504 (2012-06-22)
Stochastic simulations of coarse-grained protein models are used to investigate the propensity to form knots in early stages of protein folding. The study is carried out comparatively for two homologous carbamoyltransferases, a natively-knotted N-acetylornithine carbamoyltransferase (AOTCase) and an unknotted ornithine
James M Wilson et al.
Molecular genetics and metabolism, 105(2), 263-265 (2011-12-02)
A 66 year old woman who is a manifesting heterozygote for ornithine transcarbamylase deficiency (OTCD) presented with hepatocellular carcinoma (HCC). Fourteen years prior to this presentation she participated in a phase I gene therapy study which used an adenoviral vector
Ian E Alexander et al.
Journal of inherited metabolic disease, 35(4), 641-645 (2012-03-10)
Many metabolic diseases are compelling candidates for gene therapy, and are the subject of vigorous pre-clinical research. Successful phenotype correction in mouse models is now commonplace and research effort is increasingly being directed towards addressing the translational challenges inherent in
Gabriela Jenikova et al.
Vaccine, 29(51), 9529-9537 (2011-10-18)
Giardia lamblia is a leading protozoan cause of diarrheal disease worldwide, yet preventive medical strategies are not available. A crude veterinary vaccine has been licensed for cats and dogs, but no defined human vaccine is available. We tested the vaccine
QnAs with Arthur L. Horwich. Interview by Prashant Nair.
Arthur L Horwich
Proceedings of the National Academy of Sciences of the United States of America, 109(9), 3193-3193 (2012-02-07)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service