Skip to Content
Merck
All Photos(4)

Key Documents

N7125

Sigma-Aldrich

1-Naphthyl phosphate potassium salt

powder, >98% (NaOH, titration)

Synonym(s):

α-Naphthyl acid phosphate monopotassium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 175.00
5 G
CHF 555.00
10 G
CHF 997.00

CHF 175.00


Estimated to ship on25 April 2025



Select a Size

Change View
1 G
CHF 175.00
5 G
CHF 555.00
10 G
CHF 997.00

About This Item

Linear Formula:
C10H8O4PK
CAS Number:
Molecular Weight:
262.24
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

CHF 175.00


Estimated to ship on25 April 2025


Product Name

1-Naphthyl phosphate potassium salt, powder

Assay

>98% (NaOH, titration)

Quality Level

form

powder

color

white to faint yellow

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

[K].OP(O)(=O)Oc1cccc2ccccc12

InChI

1S/C10H9O4P.K.H/c11-15(12,13)14-10-7-3-5-8-4-1-2-6-9(8)10;;/h1-7H,(H2,11,12,13);;

InChI key

AJEYDRMDDMSWCS-UHFFFAOYSA-N

Biochem/physiol Actions

Non-specific phosphatase inhibitor. Inhibits acid, alkaline and protein phosphatases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service