Skip to Content
Merck
All Photos(1)

Key Documents

N4662

Sigma-Aldrich

Nek6, Active human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Gene A-Related Kinase 6, NIMA-Related Kinase 6, Never in Mitosis

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
CHF 469.00

CHF 469.00


Estimated to ship on04 May 2025


Request a Bulk Order

Select a Size

Change View
5 μG
CHF 469.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

CHF 469.00


Estimated to ship on04 May 2025


Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

≥80% (SDS-PAGE)

form

buffered aqueous glycerol solution

enzyme activity

≥10 units/mg protein

mol wt

protein apparent mol wt ~58 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK6(10783)

Application

Kinase activity is measured as the molar amount of phosphate incorporated into myelin basic protein, per minute per mg protein at 30 °C using a final concentration of 50 μM [32P] ATP.

Biochem/physiol Actions

Nek6 is a nuclear and cytoplasmic kinase that is required for mitotic progression of human cells. It is phosphorylated and activated during M phase of the cell cycle. Inhibition of Nek6 arrests cells in M phase and triggers apoptosis, suggesting that Nek6 is required for metaphase-anaphase transition.

Unit Definition

One unit will incorporate 1 nanomole of phosphate into myelin basic protein (MBP) per minute at pH 7.2 at 30 deg C using a final concentration of 50 μm (32p) ATP

Physical form

Solution in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, and 30% glycerol.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura O'Regan et al.
The Journal of cell biology, 209(3), 349-358 (2015-05-06)
Hsp70 proteins represent a family of chaperones that regulate cellular homeostasis and are required for cancer cell survival. However, their function and regulation in mitosis remain unknown. In this paper, we show that the major inducible cytoplasmic Hsp70 isoform, Hsp72

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service