Skip to Content
Merck
All Photos(1)

Key Documents

N214

Sigma-Aldrich

Neutrophil Activating Peptide-2 human

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

NAP-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

human

recombinant

expressed in E. coli

Assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

0.1-0.3 μg/mL ED50 (cytochalasin B treated human neutrophils)
0.1-0.5 ng/mL ED50 (BaF/3 hCXCR-2 transfected cells)

mol wt

~7.6 kDa

storage condition

avoid repeated freeze/thaw cycles (Storage in "frost-free" freezers is not recommended)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PPBP(5473)

Biochem/physiol Actions

Neutrophil Activating Peptide-2, a member of the CXC chemokine family, chemoattracts and activates neutrophils and also binds CXCR-2. NAP-2 is proteolytically processed C-terminal fragments of platelet basic protein (PBP), which is found in the α-granules of human platelets.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 0.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by the induction of myeloperoxidase release from cytochalasin B treated human neutrophils.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Proudfoot et al.
Journal of protein chemistry, 16(1), 37-49 (1997-01-01)
Connective tissue-activating peptide III (CTAP-III) and neutrophil-activating peptide-2 (NAP-2) are both derived from a common precursor, platelet basic protein (PBP), which is stored in the alpha-granules of platelets and released upon their activation. CTAP-III is an 85-residue peptide which is
N Iida et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 10(11), 1336-1345 (1996-09-01)
Leukocytes produce many biological mediators that orchestrate the subsequent cellular events during wound healing. We have identified a novel cytokine, leukocyte-derived growth factor (LDGF), which is mitogenic for connective tissue cells. Sequence analysis of the LDGF peptide revealed that it
J M Schröder et al.
Journal of immunology (Baltimore, Md. : 1950), 139(10), 3474-3483 (1987-11-15)
A novel monocyte-derived neutrophil-activating peptide (MONAP) produced by lipopolysaccharide- and phorbol myristate acetate-stimulated human peripheral blood monocytes was purified by sequential ion exchange-high performance liquid chromatography (HPLC), size exclusion HPLC, and reversed phase HPLC. Biologic activities of the purified cytokine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service