Skip to Content
Merck
All Photos(1)

Key Documents

N1390

Sigma-Aldrich

N-(2-Nitrophenylsulfenyl)-L-serine (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2O5S · C12H23N
CAS Number:
Molecular Weight:
439.57
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

C1CCC(CC1)NC2CCCCC2.OC[C@H](NSc3ccccc3[N+]([O-])=O)C(O)=O

InChI

1S/C12H23N.C9H10N2O5S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;12-5-6(9(13)14)10-17-8-4-2-1-3-7(8)11(15)16/h11-13H,1-10H2;1-4,6,10,12H,5H2,(H,13,14)/t;6-/m.0/s1

InChI key

MFUIDDKVSATADN-ZCMDIHMWSA-N

Biochem/physiol Actions

N-(2-Nitrophenylsulfenyl)-L-serine is the stable NPS derivative of L-serine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Procedure for the Preparation of Oligomer-Free N α -Fmoc Amino Acids
Nowshuddina, S., Raoa, M.N.A., and Ram Reddya, A.
Synthetic Communications, 39, 2022-2031 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service