MTCR1030
MTC™ Bio 4-Way Rack
holds 4 x 50 mL, 12 x 15 mL OR 32 x 1.5/2.0 mL, assorted colors
Synonym(s):
laboratory rack, tube holder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
assorted colors
polypropylene
feature
holds 4 x 50 mL, 12 x 15 mL OR 32 x 1.5/2.0 mL
packaging
pack of 5 ea
manufacturer/tradename
MTC™ Bio R1030
Related Categories
General description
Rack, 4 way (4x50, 12x15, 32x1.5/0.5ml), Rainbow Pack, 5/pk
Features and Benefits
- Polypropylene, withstand -80°C
- Compact and space efficient
- Autoclavable
Legal Information
MTC is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service