Key Documents
MTCM5441
MTC Bio Triangle Spreader
L × W 30 mm × 207 mm, sterile, pack of 100 ea (10 x bags 10 ea)
Synonym(s):
cell spreader
About This Item
UNSPSC Code:
41122103
NACRES:
NB.17
Pricing and availability is not currently available.
sterility
sterile
packaging
pack of 100 ea (10 x bags 10 ea)
manufacturer/tradename
MTC Bio M5441
L × W
30 mm × 207 mm
General description
Spreader Triangle, 60 x 235mm, 10 sterile peel bags of 10 spreaders, 100/pk
Features and Benefits
  • Spreads cultures and liquids evenly
  • Ideal for bacterial CFU counts
  • Smooth edges - minimizes gouging
  • Pre-sterilized - no flaming required
Legal Information
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)