MTCC4015
MTC™ Bio SureStrain™ Reducing Adaptor
for use with 15mL, 17x100mm, 12x75mm and 5mL conical for use with 15mL, 17x100mm, 12x75mm and 5mL conical tubes
Synonym(s):
cell sieve adaptor, cell strainer adaptor
Select a Size
CHF 55.40
Estimated to ship on03 April 2025
Select a Size
About This Item
CHF 55.40
Estimated to ship on03 April 2025
Recommended Products
packaging
pack of 25 ea
manufacturer/tradename
MTC™ Bio C4015
General description
Features and Benefits
- Advanced design – faster flow through
- Compatible with 6-well plates
- Interlocking and stackable
- Sterile; RNase, DNase, pyrogen and cytotoxin free
- Four 1.5mm feet for straining into flat plates and dishes
- Designed for use with 50mL tubes
Adapter for 15mL tubes included*
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service