Skip to Content
Merck
All Photos(1)

Key Documents

MSQC28

Sigma-Aldrich

SILuLite SigmaMAb Vedolizumab Monoclonal Antibody

recombinant, expressed in CHO cells

Synonym(s):

Biologic Antibody Mimic, Biotherapeutic Antibody Mimic, Monoclonal Antibody Calibrator

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
CHF 361.00

CHF 361.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
0.5 MG
CHF 361.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.24

CHF 361.00


Estimated to ship on26 May 2025


Request a Bulk Order

recombinant

expressed in CHO cells

Quality Level

Assay

≥90% (HPLC)

form

solid

suitability

suitable for mass spectrometry

shipped in

wet ice

storage temp.

−20°C

General description

SigmaMAb Vedolizumab (MSQC28) was designed to be a standard for the analysis of Vedolizumab monoclonal antibodies using mass spectrometry. An isotopically labeled version is available as SILuMab Vedolizumab (MSQC29).

SigmaMAb Vedolizumab is for research use only and is not intended for diagnostic or therapeutic use.

Sequence

SigmaMAb Vedolizumab Heavy Chain:
QVQLVQSGAEVKKPGASVKVSCKGSGYTFTSYWMHWVRQAPGQRLEWIGEIDPSESNTNYNQKFKGRVTLTVDISASTAYMELSSLRSEDTAVYYCARGGYDGWDYAIDYWGQGTLVTVSSASTKGPSVFPLAPSSKSTSGGTAALGCLVKDYFPEPVTVSWNSGALTSGVHTFPAVLQSSGLYSLSSVVTVPSSSLGTQTYICNVNHKPSNTKVDKKVEPKSCDKTHTCPPCPAPELAGAPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYVDGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKVSNKALPAPIEKTISKAKGQPREPQVYTLPPSRDELTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVMHEALHNHYTQKSLSLSPG

SigmaMAb Vedolizumab Light Chain:
DVVMTQSPLSLPVTPGEPASISCRSSQSLAKSYGNTYLSWYLQKPGQSPQLLIYGISNRFSGVPDRFSGSGSGTDFTLKISRVEAEDVGVYYCLQGTHQPYTFGQGTKVEIKRTVAAPSVFIFPPSDEQLKSGTASVVCLLNNFYPREAKVQWKVDNALQSGNSQESVTEQDSKDSTYSLSSTLTLSKADYEKHKVYACEVTHQGLSSPVTKSFNRGEC

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service