Skip to Content
Merck
All Photos(4)

Key Documents

MAK352

Sigma-Aldrich

DL-Serine Assay Kit

sufficient for 100 fluorometric tests

Synonym(s):

DL-Serine Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

relevant disease(s)

aging/geriatric diseases; neurological disorders

storage temp.

−20°C

General description

Serine is one of the 20 naturally-occurring amino acids used by all organisms in the biosynthesis of proteins. Having a single chiral center, serine can exist as either of two stereoisomers (L-Serine and D-Serine). In addition to its role as a protein building block, L-Serine serves as a precursor to many vital biomolecules, including phosphatidylserine, sphingomyelin and the amino acids glycine and cysteine. The non-proteogenic isomer D-Serine is also synthesized from L-Serine in the mammalian brain. While most D-amino acids are only present in trace amounts in mammals, D-Serine is a vital neurotransmitter that acts as a co-agonist of synaptic N-methyl-D-aspartate (NMDA) type glutamate-gated ion channels. Impaired D-Serine metabolism may contribute to several neuropsychiatric disorders, including depression, schizophrenia, epilepsy and dementia.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the Estimation of D- and L-Serine concentrations in human or animal biological fluids (plasma, serum, CSF, etc.), soft tissue homogenates (i.e. liver, brain, etc.)  and cultured cell lysates (adherent or suspension cells).

Principle

The DL-Serine Assay Kit allows for quantification of both L- and D-Serine in biological fluids and tissues. The assay is based on the conversion of L-Serine to D-Serine, which is metabolized to an intermediate product that is subsequently oxidized and reacts with a probe to form a stable fluorophore (Ex/Em = 535/587 nm). Samples may be divided and assayed simultaneously for quantification of both D-Serine and total DL-Serine. The assay is not affected by physiological concentrations of other amino acids, is high-throughput adaptable and can detect less than 1 μM Serine.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service