Skip to Content
Merck
All Photos(3)

Key Documents

MAK347

Sigma-Aldrich

Methionine Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Synonym(s):

Methionine Fluorescence Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

relevant disease(s)

cardiovascular diseases

storage temp.

−20°C

General description

Methionine (Met) is one of the twenty proteogenic amino acids encoded by the genetic code, and one of nine essential amino acids, meaning it cannot be synthesized in vivo and must be obtained from dietary sources. Met is one of two sulfur-containing amino acids (the other being cysteine) and can serve as a precursor in cysteine biosynthesis. S-adenosylmethionine, another product of Met metabolism, is an important co-factor and methyl donor in a number of biological reactions such as methylation and polyamine synthesis. Met is also an intermediate species in the synthesis of phospholipids such as lecithin, and other small precursor molecules including taurine, an aminosulfonic acid. Oxidation of Met generates methionine sulfoxide, a species whose concentration tends to increase with age and is known to cause protein mis-folding and loss-of-function of numerous enzymes. This version of the amino acid can be restored back to methionine by another enzyme, methionine sulfoxide reductase. Biologically, Met levels can affect important events such as angiogenesis, protein synthesis, and cartilage production.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the determination of methionine concentration in serum, plasma, cell cultures and tissue lysates

Principle

The Methionine Assay Kit utilizes an enzymatic mechanism by which metabolism of methionine is coupled with the stoichiometric generation of hydrogen peroxide. A probe generates fluorometric signal that can be quantified at Ex/Em: 535/587 nm. The assay shows greater than 10-fold specificity for methionine over cysteine and greater than 40-fold selectivity over other standard and non-standard amino acids. After utilization of a proprietary Sample Clean-Up Mix, the method is suitable for use in blood samples, and can detect concentrations as low as 0.5 μM, or 25 pmole Methionine.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service