Skip to Content
Merck
All Photos(1)

Key Documents

M9073

Sigma-Aldrich

Anti-Mitofusin 2 antibody,Mouse monoclonal

clone Mito-2, purified from hybridoma cell culture

Synonym(s):

Anti-CMT2A, Anti-CMT2A2, Anti-CPRP1, Anti-HSG, Anti-Hyperplasia suppressor, Anti-KIAA0214, Anti-MARF, Anti-MFN2, Anti-Mitochondrial assembly regulatory factor, Anti-Transmembrane GTPase MFN2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

Mito-2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~86 kDa

species reactivity

mouse, rat

concentration

~1.0 mg/mL

technique(s)

western blot: 2-5 μg/mL using whole extract of rat brain mitochondria

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MFN2(9927)

General description

Mitofusin-2 (MFN2) is an outer mitochondrial membrane protein.
Monoclonal Anti-Mitofusin 2 (mouse IgG1 isotype) is derived from the hybridoma Mito-2 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a fragment of human Mitofusin 2 conjugated to KLH. Mitofusin 2ismammalian homolog of the Drosophila protein fuzzy onion (Fzo). It is a transmembrane GTPases embedded in the outer membrane of mitochondria. Mitofusin 2 is broadly expressed, with highest expression in heart and skeletal muscle and is induced during myogenesis.

Application

Anti-Mitofusin 2 antibody,Mouse monoclonal has been used in western blotting.

Biochem/physiol Actions

Mitofusin 2 (Mfn2) mutant embryos have a specific and severe disruption of a layer of the placenta. Repression of Mfn2 causes morphological and functional fragmentation of the mitochondrial network into independent clusters and reduces mitochondrial membrane potential and glucose oxidation. Thus, this Mfn2-dependent regulatory mechanism is disturbed in obesity by reduced Mfn2 expression. Mutations in Mitofusin 2 cause Charcot-Marie-Tooth neuropathy type 2A, a neurological disorder that results from degeneration of axons in peripheral nerves.
Mitofusin-2 (MFN2) plays an important role as a mitochondrial fusion protein and controls many cell processes. It is mainly involved in regulating metabolic processes in the mitochondria. MFN2 prevents the formation of reactive oxygen species and controls endoplasmic reticulum stress. Mutations in the gene encoding this protein have been linked to diabetes and obesity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane topology and mitochondrial targeting of mitofusins, ubiquitous mammalian homologs of the transmembrane GTPase Fzo
Rojo M, et al.
Journal of Cell Science, 115(8), 1663-1674 (2002)
The dynamics of cardiolipin synthesis post-mitochondrial fusion
Xu FY, et al.
Biochimica et Biophysica Acta - Biomembranes, 1798(8), 1577-1585 (2010)
Short-duration swimming exercise after myocardial infarction attenuates cardiac dysfunction and regulates mitochondrial quality control in aged mice
Zhao D, et al.
Oxidative Medicine and Cellular Longevity, 2018 (2018)
Mutations in the mitochondrial GTPase mitofusin 2 cause Charcot-Marie-Tooth neuropathy type 2A
Zuchner S, et al.
Nature Genetics, 36(5), 449-449 (2004)
Mitofusin-2 determines mitochondrial network architecture and mitochondrial metabolism A novel regulatory mechanism altered in obesity
Bach D, et al.
The Journal of Biological Chemistry, 278(19), 17190-17197 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service