Skip to Content
Merck
All Photos(1)

Key Documents

M5026

Sigma-Aldrich

Methyl 4-O-β-D-galactopyranosyl-β-D-glucopyranoside

Synonym(s):

β-D-Gal-[1→4]-β-D-Glc-1→OMe, Methyl β-lactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24O11
CAS Number:
Molecular Weight:
356.32
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

[H][C@]1(O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)O[C@H]2[C@H](O)[C@@H](O)[C@H](OC)O[C@@H]2CO

InChI

1S/C13H24O11/c1-21-12-10(20)8(18)11(5(3-15)23-12)24-13-9(19)7(17)6(16)4(2-14)22-13/h4-20H,2-3H2,1H3/t4-,5-,6+,7+,8-,9-,10-,11-,12-,13+/m1/s1

InChI key

FHNIYFZSHCGBPP-ABBMIVAOSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Bock et al.
Carbohydrate research, 202, 131-149 (1990-07-15)
The 2'-,4'-, and 6'-deoxy derivatives of methyl beta-lactoside have been synthesised by deoxygenation at positions 2', 4', and 6', and the 3'-deoxy derivative was obtained by a glycosylation reaction. The 2'-O-methyl, 2'-O-benzyl, 2'-amino-2'-deoxy, and 1'-deuterio derivatives have been synthesized also.
P Fernández et al.
Carbohydrate research, 248, 15-36 (1993-10-04)
The solution conformations of all the possible monomethyl ethers of methyl beta-lactoside have been analysed using molecular mechanics and dynamics calculations and nuclear magnetic resonance data (variable temperature and NOE experiments). The overall shape of all the compounds studied is
D Solís et al.
European journal of biochemistry, 214(3), 677-683 (1993-06-15)
The binding of O-methyl and fluorodeoxy derivatives of methyl beta-lactoside to the Ricinus communis toxin (RCA60) and agglutinin (RCA120) was studied in order to determine the donor/acceptor relationships of the hydrogen bonds between the hydroxyl groups of methyl beta-lactoside and
Xiaosong Hu et al.
Acta crystallographica. Section C, Crystal structure communications, 66(Pt 2), o67-o70 (2010-02-04)
Methyl beta-D-galactopyranosyl-(1-->4)-alpha-D-mannopyranoside methanol 0.375-solvate, C(13)H(24)O(11).0.375CH(3)OH, (I), was crystallized from a methanol-ethanol solvent system in a glycosidic linkage conformation, with varphi' (O5(Gal)-C1(Gal)-O1(Gal)-C4(Man)) = -68.2 (3) degrees and psi' (C1(Gal)-O1(Gal)-C4(Man)-C5(Man)) = -123.9 (2) degrees , where the ring is defined by atoms
P Fernández et al.
Carbohydrate research, 256(2), 223-244 (1994-04-01)
Syntheses of the 3-aminodeoxy (4), 3-deoxy-3-methyl (5), and 3-epi (6) derivatives of methyl beta-lactoside (1) have been achieved from 1 in a straightforward way, and their solution conformations in water and dimethyl sulfoxide analysed through molecular mechanics and dynamics calculations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service