Skip to Content
Merck
All Photos(1)

Key Documents

M2692

Sigma-Aldrich

MK-886 sodium salt hydrate

>98% (HPLC)

Synonym(s):

3-[3-tert-Butylthio-1-(4-chlorobenzyl)-5-isopropyl-1H-indol-2-yl]-2,2-dimethylpropionic acid, sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H33NO2ClSNa · xH2O
CAS Number:
Molecular Weight:
494.06 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>98% (HPLC)

color

white

solubility

DMSO: 32 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

O.[Na+].CC(C)c1ccc2n(Cc3ccc(Cl)cc3)c(CC(C)(C)C([O-])=O)c(SC(C)(C)C)c2c1

InChI

1S/C27H34ClNO2S.Na.H2O/c1-17(2)19-10-13-22-21(14-19)24(32-26(3,4)5)23(15-27(6,7)25(30)31)29(22)16-18-8-11-20(28)12-9-18;;/h8-14,17H,15-16H2,1-7H3,(H,30,31);;1H2/q;+1;/p-1

InChI key

APMAGPLTTCSTMM-UHFFFAOYSA-M

Biochem/physiol Actions

Potent and specific inhibitor of leukotriene biosynthesis.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Hsuan Wang et al.
Brain research bulletin, 79(3-4), 169-176 (2009-05-12)
Molecules involved in self-protection of neurons against glucose/oxygen/serum deprivation (GOSD) were investigated. Trypan blue dye exclusion assay, Western blotting, ELISA, cytokine antibody array and chemical blocking assay were applied in the study. Results showed that early induction (at 6h of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service