Skip to Content
Merck
All Photos(1)

Key Documents

L9631

Sigma-Aldrich

Myristyl dodecanoate

≥99%

Synonym(s):

Lauric acid myristyl ester, Myristyl laurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H52O2
CAS Number:
Molecular Weight:
396.69
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥99%

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCOC(=O)CCCCCCCCCCC

InChI

1S/C26H52O2/c1-3-5-7-9-11-13-14-15-17-19-21-23-25-28-26(27)24-22-20-18-16-12-10-8-6-4-2/h3-25H2,1-2H3

InChI key

FNMPODAQERUMDD-UHFFFAOYSA-N

Biochem/physiol Actions

Myristyl dodecanoate (Myristyl laurate) is a laurate medium-chain unsaturated fatty acid ester compound that may be used as a reference in the analysis of wax esters of lauric acid.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural characterization of wax esters by electron ionization mass spectrometry.
Urbanova K, Vrkoslav V, Valterova I, et al.
Journal of Lipid Research, 53, 2004-2213 (2012)
M C Pérez-Camino et al.
Journal of chromatography. A, 983(1-2), 283-288 (2003-02-06)
A procedure for the simultaneous determination of long-chain aliphatic aldehydes, and aliphatic and triterpenic waxes in virgin olive oils is described. A fraction containing these compounds was isolated from the oil using solid-phase extraction on silica-gel cartridges. The fraction was
Vladimír Vrkoslav et al.
Journal of chromatography. A, 1217(25), 4184-4194 (2010-01-19)
High chromatographic resolution of wax esters (WEs) was achieved by non-aqueous reversed-phase liquid chromatography on a Nova-Pak C18 column by optimising the acetonitrile/ethyl acetate mobile phase gradient. The retention behaviour of WEs was studied in this chromatographic system. The WEs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service