Skip to Content
Merck
All Photos(1)

Key Documents

L9007

Sigma-Aldrich

Luciferin 6′-ethyl ether sodium salt

Synonym(s):

4,5-Dihydro-2-(6-ethoxy-2-benzothiazolyl)-4-thiazolinecarboxylic acid sodium salt, 6′-O-Ethylluciferin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C13H12N2O3S2
CAS Number:
Molecular Weight:
330.36
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥99%

form

powder

storage temp.

−20°C

SMILES string

[Na+].CCOc1ccc2nc(sc2c1)C3=N[C@H](CS3)C([O-])=O

InChI

1S/C13H12N2O3S2.Na/c1-2-18-7-3-4-8-10(5-7)20-12(14-8)11-15-9(6-19-11)13(16)17;/h3-5,9H,2,6H2,1H3,(H,16,17);/q;+1/p-1/t9-;/m1./s1

InChI key

PYKJITLAHMFBSA-SBSPUUFOSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service