Key Documents
L5664
Sigma-Aldrich
Laser Tough-Spots®
diam. 3/8 in., orange
Synonym(s):
solvent resistant labels, temperature resistant labels
Select a Size
1 PKG
CHF 77.50
CHF 77.50
Check Cart for Availability
Select a Size
Change View
1 PKG
CHF 77.50
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
CHF 77.50
Check Cart for Availability
material
orange
polyolefin
packaging
pack of 3840 ea (192 labels per sheet)
manufacturer/tradename
Diversified Biotech SPOT-1300
diam.
3/8 in.
Related Categories
General description
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Legal Information
Tough-Spots is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)