Skip to Content
Merck
All Photos(1)

Key Documents

L5646

Sigma-Aldrich

Trizma® dodecyl sulfate

Synonym(s):

Tris lauryl sulfate, Tris(hydroxymethyl)aminomethane lauryl sulfate, Trizma® lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · CH3(CH2)11OSO3H
CAS Number:
Molecular Weight:
387.53
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

anionic

SMILES string

NC(CO)(CO)CO.CCCCCCCCCCCCOS(O)(=O)=O

Looking for similar products? Visit Product Comparison Guide

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service