Skip to Content
Merck
All Photos(1)

Key Documents

L5538

Sigma-Aldrich

Leu-Ser-Lys-Leu-NH2

≥95% (HPLC), suitable for mass spectrometry

Synonym(s):

LSKL-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H42N6O5
Molecular Weight:
458.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Product Name

Leu-Ser-Lys-Leu-NH2, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

white

storage temp.

−20°C

SMILES string

CC(C)C[C@H](N)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(N)=O

InChI

1S/C21H42N6O5/c1-12(2)9-14(23)19(30)27-17(11-28)21(32)25-15(7-5-6-8-22)20(31)26-16(18(24)29)10-13(3)4/h12-17,28H,5-11,22-23H2,1-4H3,(H2,24,29)(H,25,32)(H,26,31)(H,27,30)/t14-,15-,16-,17-/m0/s1

InChI key

YOKXDNNIFSAXBY-QAETUUGQSA-N

Amino Acid Sequence

Leu-Ser-Lys-Leu-NH2

Biochem/physiol Actions

Leu-Ser-Lys-Leu-NH2 (LSKL-NH2) is found at the C-terminal of the neurotoxic wasp venom α-pompilidotoxin (α-PMTX). A similar peptide Leu-Ser-Lys-Leu (LSKL) is a used to study the activation of TGF-β1 via thrombospondin.
Used to study the activation of TGF-β1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yurong Zhang et al.
Brain research bulletin, 149, 106-119 (2019-04-22)
Epilepsy is accompanied by angiogenesis and blood-brain barrier (BBB) disruption. The transforming growth factor-β1 (TGF-β1)/phosphorylated small mothers against decapentaplegic 2 and 3 (pSmad2/3)/vascular endothelial growth factor (VEGF) pathway, activated by thrombospondin-1 (TSP-1), which is further regulated by Y type P2
Xi-sheng Xie et al.
Archives of pharmacal research, 33(2), 275-284 (2010-03-03)
The effects of LSKL, the peptide antagonist of thrombospondin-1 (TSP-1), on renal interstitial fibrosis in rats subjected to unilateral ureteral obstruction (UUO) were investigated. Rats were divided randomly into three groups (n = 20 each): UUO group, sham-operation group and
Emanuele Schiavon et al.
The FEBS journal, 277(4), 918-930 (2010-01-12)
Pompilidotoxins (PMTXs, alpha and beta) are small peptides consisting of 13 amino acids purified from the venom of the solitary wasps Anoplius samariensis (alpha-PMTX) and Batozonellus maculifrons (beta-PMTX). They are known to facilitate synaptic transmission in the lobster neuromuscular junction
K Konno et al.
Biochemical and biophysical research communications, 250(3), 612-616 (1998-10-24)
Novel peptide neurotoxins, alpha- and beta-pompilidotoxins (alpha- and beta-PMTXs), were purified from the venoms of the solitary wasps Anoplius samariensis and Batozonellus maculifrons. Their structures were analyzed mostly by MALDI-TOF-MS, which were corroborated by solid-phase synthesis. alpha-PMTX, with 13 amino

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service