Skip to Content
Merck
All Photos(1)

Key Documents

L5417

Sigma-Aldrich

Anti-phospho-LAT (pTyr191) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

western blot: 0.1-1.0 μg/mL using stimulated Jurkat E6.1 cells

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LAT(27040)
mouse ... Lat(16797)
rat ... Lat(81511)

Immunogen

synthetic phosphopeptide derived from the region of human LAT that contains tyrosine 191.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 1.0 mg/mL BSA (IgG and protease free) and 0.05% sodium azide.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service