Skip to Content
Merck
All Photos(1)

Key Documents

L5386

Sigma-Aldrich

[des-Gly10, D-Trp6]-LH-RH ethylamide

≥97% (HPLC)

Synonym(s):

Deslorelin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H83N17O12
CAS Number:
Molecular Weight:
1282.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CCNC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@H](CO)NC(=O)[C@H](Cc5c[nH]c6ccccc56)NC(=O)[C@H](Cc7c[nH]cn7)NC(=O)[C@@H]8CCC(=O)N8

InChI

1S/C64H83N17O12/c1-4-68-62(92)53-16-10-24-81(53)63(93)46(15-9-23-69-64(65)66)74-56(86)47(25-35(2)3)75-58(88)49(27-37-30-70-43-13-7-5-11-41(37)43)77-57(87)48(26-36-17-19-40(83)20-18-36)76-61(91)52(33-82)80-59(89)50(28-38-31-71-44-14-8-6-12-42(38)44)78-60(90)51(29-39-32-67-34-72-39)79-55(85)45-21-22-54(84)73-45/h5-8,11-14,17-20,30-32,34-35,45-53,70-71,82-83H,4,9-10,15-16,21-29,33H2,1-3H3,(H,67,72)(H,68,92)(H,73,84)(H,74,86)(H,75,88)(H,76,91)(H,77,87)(H,78,90)(H,79,85)(H,80,89)(H4,65,66,69)/t45-,46-,47-,48-,49+,50-,51-,52-,53-/m0/s1

InChI key

GJKXGJCSJWBJEZ-XRSSZCMZSA-N

Amino Acid Sequence

pGlu-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-NHEt

Biochem/physiol Actions

Long-acting LH-RH agonist.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S A Colgan et al.
Australian veterinary journal, 97(12), 515-523 (2019-11-21)
A large-scale capture method was developed to enable sterilisation of a macropod population in western Sydney from 2005 to 2018. Until March 2007, free ranging eastern grey kangaroos and red kangaroos were herded into purpose-built 15 m diameter capture yards
Robert M Kirberger et al.
BMC veterinary research, 13(1), 312-312 (2017-11-08)
Cheetahs in captivity have a high prevalence of chronic renal diseases. We ultrasonographically evaluated the renal volumes, a variety of renal dimensions, interarcuate artery resistive indices (RI) as well as aortic diameters and the length of the ventral aspect of the
Wojciech Niżański et al.
Animals : an open access journal from MDPI, 10(12) (2020-12-17)
This article presents B-mode and color Doppler imaging of the prostate and testes in dogs suffering from benign prostate hyperplasia (BPH), and receiving deslorelin acetate (SuprelorinTM) or osaterone acetate (YpozaneTM). The study was planned as a controlled clinical trial, dogs
L David Mech et al.
PloS one, 12(9), e0184342-e0184342 (2017-09-02)
Following the production of western gray wolf (Canis lupus) x western coyote (Canis latrans) hybrids via artificial insemination (AI), the present article documents that the hybrids survived in captivity for at least 4 years and successfully bred with each other.
Marta Cecchetto et al.
Journal of feline medicine and surgery, 19(12), 1224-1230 (2017-02-15)
Objectives The purpose of this study was to assess efficacy of deslorelin, a gonadotropin-releasing hormone (GnRH) agonist marketed in Europe for the control of male dog reproduction, for the postponement of puberty in queens. Methods Nine prepubertal queens aged 3-9

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service