Skip to Content
Merck
All Photos(1)

Key Documents

L4295

Sigma-Aldrich

LDN-91946

≥98% (HPLC)

Synonym(s):

3-Amino-2-benzoyl-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2O4S
CAS Number:
Molecular Weight:
314.32
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

brown

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

Biochem/physiol Actions

LDN-91946 is a potent, selective and uncompetitive UCH-L1 inhibitor that targets the enzyme-substrate complex, but not the free enzyme.
LDN-91946 is a potent, selective and uncompetitive UCH-L1 inhibitor that targets the enzyme-substrate complex, but not the free enzyme. Ubiquitin C-terminal hydrolase L1 (UCH-L1) is a deubiquitinating enzyme. It is one of the most abundant proteins in the brain. UCKL1 plays a key role in processing polyubiquitin and ubiquitylated proteolytic peptides. The enzyme has been linked to Parkinson disease and lung cancer. LDN-91946 inhibits UCH-L1-catalyzed substrate hydrolysis in an uncompetitive manner by targeting the enzyme-substrate complex, but not the free enzyme. The compound is quite specific and is exhibiting little activity against UCH-L3, TGase 2, Papain, or Caspase-3 even at concentrations as high as 20 μM. Compound was recently launched by Calbiochem. LDN-57444 is acts as competitive inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service