Skip to Content
Merck
All Photos(1)

Key Documents

L3759

Sigma-Aldrich

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia)

biotin conjugate, lyophilized powder

Synonym(s):

Bandeirea simplicifolia agglutinin, BS-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

conjugate

biotin conjugate

form

lyophilized powder

potency

<250 μg per mL agglutination activity (using human blood group A erythrocytes.)

composition

Protein, ~85% E1%/280

extent of labeling

~5 mol biotin per mol protein

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia) has been used:
  • for highlighting murine endothelial cells
  • to identify fetal capillaries in endothelial immunohistochemical staining, conjugated with biotin
  • to detect tumor total vessels

Biochem/physiol Actions

BS-I has a major affinity for terminal α-D-galactosyl residues with a secondary affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Other Notes

BS-I is a tetrameric lectin consisting of two types of subunits designated A and B. There are five BS-I isolectins with different subunit composition: BSI-B4, BSI-AB3, BSI-A2B2, BSI-A3B and BSI-A4. BSI-B4 is blood group B specific and has an exclusive affinity for terminal α-D-galactosyl residues, whereas BSI-A4 has blood group A specificity and has a major affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Physical form

Contains sodium citrate

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions of a 1 mg/mL solution using phosphate buffered saline, pH 6.8, containing, for each lectin, calcium, magnesium, and manganese at different concentrations. This activity is the lowest concentration to agglutinate a 2% suspension of appropriate erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Schaad et al.
Scientific reports, 7, 41842-41842 (2017-02-09)
A detailed vascular visualization and adequate quantification is essential for the proper assessment of novel angiomodulating strategies. Here, we introduce an ex vivo micro-computed tomography (microCT)-based imaging approach for the 3D visualization of the entire vasculature down to the capillary
Intravitreal and subconjunctival melphalan for retinoblastoma in transgenic mice
Shah NV, et al.
Journal of Ophthalmology, 2014 (2014)
Yun Sook Kim et al.
The Journal of comparative neurology, 518(5), 687-698 (2009-12-25)
Transient receptor potential ankyrin 1 (TRPA1), responding to noxious cold and pungent compounds, is implicated in the mediation of nociception, but little is known about the processing of the TRPA1-mediated nociceptive information within the trigeminal sensory nuclei (TSN) and the
Merle M Krebber et al.
Journal of hypertension, 38(3), 489-503 (2019-10-28)
Left ventricular (LV) hypertrophy is the most common cardiac alteration in patients with chronic kidney disease (CKD). Normalization of hypertension in CKD patients receiving a healthy kidney allograft often reverses LV hypertrophy, but effects on LV fibrosis remain unclear. To
Suppressive effect of 2, 3, 7, 8-tetrachlorodibenzo-p-dioxin on vascular remodeling that takes place in the normal labyrinth zone of rat placenta during late gestation
Ishimura R, et al.
Toxicological Sciences, 91(1), 265-274 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service