Skip to Content
Merck
All Photos(1)

Key Documents

L2545

Sigma-Aldrich

LY225910

≥98% (HPLC)

Synonym(s):

2-[2-(5-Bromo-1H-indol-3-yl)ethyl]-3-[3-(1-methylethoxy )phenyl]-4-(3H)-quinazolinone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CHF 331.00

CHF 331.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
CHF 331.00

About This Item

Empirical Formula (Hill Notation):
C27H24BrN3O2
CAS Number:
Molecular Weight:
502.40
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

CHF 331.00


Estimated to ship on25 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: >20 mg/mL

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

CC(C)Oc1cccc(c1)N2C(CCc3c[nH]c4ccc(Br)cc34)=Nc5ccccc5C2=O

InChI

1S/C27H24BrN3O2/c1-17(2)33-21-7-5-6-20(15-21)31-26(30-25-9-4-3-8-22(25)27(31)32)13-10-18-16-29-24-12-11-19(28)14-23(18)24/h3-9,11-12,14-17,29H,10,13H2,1-2H3

InChI key

KUECXUACQOYKNB-UHFFFAOYSA-N

Biochem/physiol Actions

Cholecystokinin is expressed in the gastrointestinal tract and the central nervous system. Cholecystokinin receptor type 2 (CCK2) is a GPCR that is highly expressed in brain and spinal cord. CCK2 is implicated in many brain processes, including mood, anxiety, and pain, through its modulation of GABA neurotransmission.

LY225910 is a potent, selective CCK2 antagonist. LY225910 blocks agonism of CCK2 by CCK-8S, the peptide agonist. LY225910 has been measured in multiple systems, including GABA efflux from cortical cultures, depolarization of spinal cord neurons (via potassium conductance), modulation of excitatory postsynaptic potentials (EPSPs) in nucleus accumbens slices, and enhancement of morphine analgesia.
LY 225910 is a potent, selective CCK2 antagonist.

Features and Benefits

This compound is featured on the Cholecystokinin and Gastrin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

638.6 - 762.1 °F

Flash Point(C)

337 - 405.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service