Skip to Content
Merck
All Photos(1)

Key Documents

L1170

Sigma-Aldrich

LY-487379 hydrochloride

≥98% (HPLC)

Synonym(s):

N-(4-(2-Methoxyphenoxy)phenyl)-N-(2,2,2- trifluoroethylsulfonyl)pyrid-3-ylmethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19F3N2O4SHCl
CAS Number:
Molecular Weight:
488.91
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white

solubility

DMSO: >20 mg/mL

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

Cl.COc1ccccc1Oc2ccc(cc2)N(Cc3cccnc3)S(=O)(=O)CC(F)(F)F

InChI

1S/C21H19F3N2O4S.ClH/c1-29-19-6-2-3-7-20(19)30-18-10-8-17(9-11-18)26(14-16-5-4-12-25-13-16)31(27,28)15-21(22,23)24;/h2-13H,14-15H2,1H3;1H

InChI key

LPWFRDWTOKLHJC-UHFFFAOYSA-N

Biochem/physiol Actions

LY-487379 is a positive allosteric modulator of the metabotropic glutamate receptor mGluR2.
LY-487379 is a positive allosteric modulator of the metabotropic glutamate receptor mGluR2. EC50 = 1.7 μM. LY-487379 shows no intrinsic agonist or antagonist activity at hmGluR2 and has no activity at human mGluR3. LY-487379 markedly potentiates glutamate-stimulated [35S]GTPγS binding in a concentration-dependent manner at hmGluR2, shifting the glutamate dose-response curve leftward by 3-fold and increasing the maximum levels of [35S]GTPγS stimulation. LY-487379 has anxiolytic effects in fear-potentiated startle. This activity profile is useful in all symptom domains of schizophrenia.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suli Chen et al.
Molecular cell, 74(1), 118-131 (2019-03-02)
Alternative polyadenylation (APA) produces mRNA isoforms with different 3' UTR lengths. Previous studies indicated that 3' end processing and mRNA export are intertwined in gene regulation. Here, we show that mRNA export factors generally facilitate usage of distal cleavage and
Lilian Phu et al.
Molecular cell, 77(5), 1107-1123 (2020-03-07)
Mitochondria import nearly their entire proteome from the cytoplasm by translocating precursor proteins through the translocase of the outer membrane (TOM) complex. Here, we show dynamic regulation of mitochondrial import by the ubiquitin system. Acute pharmacological inhibition or genetic ablation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service