Skip to Content
Merck
All Photos(1)

Key Documents

K4639

Sigma-Aldrich

Anti-Potassium Channel Kv1.7 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-KCNA7, Anti-Voltage gated K+ channel KV1.7

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
CHF 596.00

CHF 596.00


Estimated to ship on28 April 2025


Request a Bulk Order

Select a Size

Change View
200 μL
CHF 596.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 596.00


Estimated to ship on28 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

should not react with

human

technique(s)

western blot: 1:200 using heart membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Kcna7(16495)

Immunogen

peptideTTRKAQEIHGKAPG (C) corresponding to residues 2-15 of mouse Kv1.7.

Target description

Kv1.7 voltage-gated K+ channel is a member of the Shaker family of K+ channels that includes eight members (Kv1.1- Kv1.8). The biophysical and pharmacological properties of the Kv1.7 channel closely resemble that of the ultra-rapidly activating delayed rectifier (IKur) in cardiac tissue. This current plays a central role in cardiac atria repolarization that was largely believed to correspond to the activity of the Kv1.5 channel.

Physical form

Lyophilized powder from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.025% sodium azide.

Reconstitution

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein such as BSA (1-3%).

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service