Skip to Content
Merck
All Photos(1)

Documents

K4263

Sigma-Aldrich

Anti-Kaiso antibody ,Mouse monoclonal

clone 6F, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

6F, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat, canine, chicken

technique(s)

microarray: suitable
western blot: 1-2 μg/mL using CSMLO (mouse mammary adenocarcinoma) cell extracts

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-Kaiso (mouse IgG1 isotype) is derived from the 6F hybridoma produced by the fusion of mouse myeloma cells and splenocytes from A/J mice. Kaiso is a DNA-binding protein and a member of the broad complex, tramtrak, bric a brac/ pox zinc finger (BTB/POZ) family of transcription factors with zinc finger motifs. This protein contains three carboxy-terminal zinc-finger motifs of the Kruppel-like C2H2-DNA binding type zinc fingers and a poxvirus and zinc finger (POZ) domain in its extreme N-terminus, which is highly hydrophobic. The zinc finger and BTB domain containing 33 (ZBTB33) gene is located on the human chromosome at Xq24.

Specificity

Monoclonal Anti-Kaiso reacts specifically with human, rat, mouse, canine, and chicken Kaiso (approx.105 kDa).

Immunogen

recombinant purified mouse Kaiso protein (amino acids 1-499).

Application

Anti-Kaiso antibody, Mouse monoclonal may be used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry

Biochem/physiol Actions

Kaiso is a transcriptional repressor that recognizes specifically the DNA sequence TCCTGCNA or two symmetrically methylated CpG dinucleotides. This protein binds to the cell adhesion cofactor p120 ctn which signifies its role in the p120 ctn action and signaling.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliet M Daniel et al.
Nucleic acids research, 30(13), 2911-2919 (2002-06-28)
The p120(ctn)-binding partner Kaiso is a new member of the POZ-zinc finger family of transcription factors implicated in development and cancer. To understand the role of Kaiso in gene regulation and p120(ctn)-mediated signaling and adhesion, we sought to identify Kaiso-specific
Filip Majer et al.
American journal of medical genetics. Part A, 182(1), 219-223 (2019-11-16)
Cullin 4B (CUL4B), lysosomal-associated membrane protein Type 2 (LAMP2), ATP1B4, TMEM255A, and ZBTB33 are neighboring genes on Xq24. Mutations in CUL4B result in Cabezas syndrome (CS). Male CS patients present with dysmorphic, neuropsychiatric, genitourinary, and endocrine abnormalities. Heterozygous CS females
J M Daniel et al.
Molecular and cellular biology, 19(5), 3614-3623 (1999-04-17)
p120(ctn) is an Armadillo repeat domain protein with structural similarity to the cell adhesion cofactors beta-catenin and plakoglobin. All three proteins interact directly with the cytoplasmic domain of the transmembrane cell adhesion molecule E-cadherin; beta-catenin and plakoglobin bind a carboxy-terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service