Skip to Content
Merck
All Photos(1)

Documents

IST102078LS

ClearASeal Weld heat sealable film

LabSheet, W × L 78 mm × 125 mm, pack of 100 sheets

Synonym(s):

non-pierceable film, permanent plate seal, PCR storage, qPCR storage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.15

sterility

non-sterile

packaging

pack of 100 sheets

manufacturer/tradename

IST Scientific IST-102-078LS

technique(s)

PCR: suitable
qPCR: suitable

W × L

78 mm × 125 mm

suitability

suitable for PCR
suitable for fluorescence

Looking for similar products? Visit Product Comparison Guide

General description

Optically clear polymer heat sealing film used to form a permanent seal to polypropylene plates, enabling both low and very high temperature uses, including low temperature storage and high temperature incubations. Suitable for PCR/qPCR, even without the use of a pressurized heated lid, and 100% effective when used in water bath thermal cyclers. The permanent nature of the seal renders it suitable for the storage and disposal of hazardous material.

Application

  • Permanent seal
  • Difficult to pierce
  • Seal integrity range: -80°C to 110°C
  • DMSO and solvent resistant
  • Autoclavable (121°C)

Features and Benefits

  • Permanent seal
  • Difficult to pierce
  • Seal integrity range: -80°C to 110°C
  • DMSO and solvent resistant
  • Autoclavable (121°C)

Legal Information

ClearASeal Peel is a trademark of IST Scientific
LabSheet is a trademark of IST Scientific

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service