Skip to Content
Merck
All Photos(1)

Key Documents

I2786

Sigma-Aldrich

IL-6 human

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Interleukin-6 human, B cell stimulating factor (BSF-2), IL-6, IL6, hepatocyte stimulating factor (HSF), interferon b2 (IFNb2), interleukin hybridoma/plasmacytoma-1 (IL-HP1), monocyte derived human B cell growth factor, plasmacytoma growth factor (PCT-GF)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Interleukin-6 human, IL-6, recombinant, expressed in E. coli, Carrier Free

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

0.2-2.0 ng/mL Biological Activity EC50

quality

Carrier Free

mol wt

26 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

≤1 EU/μg protein Endotoxin Level

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL6(3569)

General description

Interleukin-6 (IL-6) is a multi-functional 26 kDa protein originally discovered in the medium of RNA-stimulated fibroblastoid cells1. IL-6 appears to be directly
involved in the immune responses that occur after infection and cellular injury, and it may prove to be as important as IL-1 and TNF-a in regulating the acute phase response2,3.

IL-6 is reportedly produced by fibroblasts, activated T cells, activated monocytes or macrophages, and endothelial cells. It acts upon a variety of cells including fibroblasts, myeloid progenitor cells, T cells, B cells, and hepatocytes. IL-6 induces multiple effects as indicated by its numerous synonyms4. In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes5. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors6.This product is lyophilized from a 0.2 mm-filtered solution of phosphate buffered saline, pH 7.4, containing a stabilizer.

The product was prepared from raw materials verified to be carrier free.

Application

Human interleukin-6 (IL-6) has been used to study the effects of IL-6, along with IL-8 and IL-1β, on the clot structure of healthy blood.[1]

Biochem/physiol Actions

IL-6 is a multifunctional protein originally discovered in the media of cells stimulated with double stranded RNA. IL-6 appears to be directly involved in the responses that occur after infection and injury and may prove to be as important as IL-1 and TNF-α in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages, and endothelial cells. It acts upon a variety of cells, including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. IL-6 induces multiple effects, as indicated by its numerous synonyms: plasmacytoma growth factor (PCT-GF), interferon-β-2 (IFN-β2), monocyte derived human B cell growth factor, B cell stimulating factor (BSF-2), hepatocyte stimulating factor (HSF), Interleukin Hybridoma/Plasmacytoma-1 (IL-HP1). In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 also potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors.
Interleukin-6 is a multifunctional protein that is involved in the responses that occur after infection and injury and may prove to be as important as IL-1 and TNF-α in regulating the acute phase response. IL-6 induces multiple effects. It interacts with IL-2 in the proliferation of T lymphocytes, and potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tadamitsu Kishimoto
Clinical reviews in allergy & immunology, 28(3), 177-186 (2005-09-01)
In the late 1960s, the essential role of T-cells in antibody production was reported. This suggested to us that certain molecules should be released from T-cells for the stimulation of B-cells. We discovered activities in the culture supernatant of T-cells
Simultaneous presence of hypercoagulation and increased clot lysis time due to IL-1beta, IL-6 and IL-8
Janette B, et al.
Cytokine (2018)
Interleukin-6: an overview.
J Van Snick
Annual review of immunology, 8, 253-278 (1990-01-01)
A new interleukin with pleiotropic activities.
T Kishimoto et al.
BioEssays : news and reviews in molecular, cellular and developmental biology, 9(1), 11-15 (1988-07-01)
J Van Snick et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(24), 9679-9683 (1986-12-01)
A T-cell-derived lymphokine was identified by its ability to support the growth of a subset of B-cell hybridomas. Hybrids that failed to survive in the absence of this molecule represented a major proportion of rat-mouse hybridomas but were very rare

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service