Skip to Content
Merck
All Photos(1)

Key Documents

I0510

Sigma-Aldrich

D-myo-Inositol 1,4-bisphosphate potassium salt

from bovine brain, ≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12K2O12P2
CAS Number:
Molecular Weight:
416.30
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

biological source

bovine brain

Assay

≥98% (TLC)

storage temp.

−20°C

SMILES string

[K+].O[C@@H]1[C@@H](O)[C@H](OP(O)([O-])=O)[C@H](O)[C@H](O)[C@H]1OP(O)(O)=O

InChI

1S/C6H14O12P2.K/c7-1-2(8)6(18-20(14,15)16)4(10)3(9)5(1)17-19(11,12)13;/h1-10H,(H2,11,12,13)(H2,14,15,16);/q;+1/p-1/t1-,2-,3-,4+,5+,6+;/m1./s1

InChI key

UEZPJWQBOCTMMK-HTBPUPAMSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phosphoinostol broken down from glycerophosphoinositols

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I M Bird et al.
Molecular and cellular endocrinology, 66(2), 215-229 (1989-10-01)
The optimisation of a reaction for the conversion of glycerophosphoinositols to phosphoinositols is described. This reaction has been used in a scheme, described in detail, for the formation of D-myo-inositol 1,4-bisphosphate and D-myo-inositol 1,4,5-trisphosphate in mg quantities from a readily
A Emilsson et al.
The Journal of biological chemistry, 259(5), 3111-3116 (1984-03-10)
The inositol phospholipids of peritoneal macrophages were prelabeled with [3H]inositol to enable studies on the enzymatic mechanisms of stimulus-induced phosphatidylinositol breakdown. Ionophore A23187 induced a rapid breakdown of phosphatidylinositol in the presence of Ca2+ with 25% loss occurring within 5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service