Skip to Content
Merck
All Photos(6)

Key Documents

HPA027302

Sigma-Aldrich

Anti-CAMSAP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3

Synonym(s):

Anti-CAMSAP1L1, Anti-KIAA1078

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 556.00

CHF 556.00


Check Cart for Availability


Select a Size

Change View
100 μL
CHF 556.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

CHF 556.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KLVTERDLHKKPIQMSAHLAMIDTLMMAYTVEMVSIEKVIACAQQYSAFFQATDLPYDIEDAVMYWINKVNEHLKDIMEQEQKLKEHHTVEAPGG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Calmodulin regulated spectrin associated protein family member 2 (CAMSAP2) is a cytoskeletal protein. The gene encoding it is localized on human chromosome 1q32.1.

Immunogen

Calmodulin-regulated spectrin-associated protein 1-like protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Calmodulin regulated spectrin associated protein family member 2 (CAMSAP2) associates with the minus-end of microtubules and plays a role in their organization in neurons. It also functions in dendrite branch formation, neuronal polarity and axon specification. CAMSAP2 may have a role in symptomatic epilepsy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76667

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congcong Dong et al.
Journal of cell science, 130(10), 1709-1715 (2017-04-08)
The epithelium has an apico-basal axis polarity that plays an important role in absorption, excretion and other physiological functions. In epithelial cells, a substantial number of non-centrosomal microtubules (MTs) are scattered in the cytoplasm with an apico-basal polarity and reorientate
Youling Guo et al.
Human molecular genetics, 21(5), 1184-1189 (2011-11-26)
In the majority of patients, epilepsy is a complex disorder with multiple susceptibility genes interacting with environmental factors. However, we understand little about its genetic risks. Here, we report the first genome-wide association study (GWAS) to identify common susceptibility variants
Shuai Zhang et al.
Neuroscience letters, 556, 146-151 (2013-10-24)
In a recent genome-wide association study (GWAS) of symptomatic epilepsy in the Chinese population, the most significant single nucleotide polymorphism (SNP) allele was rs2292096 [G] (P=1.0×10(-8), odds ratio [OR]=0.63), in the CAMSAP1L1 gene (also known as CAMSAP2). Here, we report
Kah Wai Yau et al.
Neuron, 82(5), 1058-1073 (2014-06-09)
In neurons, most microtubules are not associated with a central microtubule-organizing center (MTOC), and therefore, both the minus and plus-ends of these non-centrosomal microtubules are found throughout the cell. Microtubule plus-ends are well established as dynamic regulatory sites in numerous
Jieli Wei et al.
FEBS letters, 591(16), 2379-2393 (2017-07-21)
Microtubules (MTs) play essential roles in many steps of autophagy, an important degradation pathway in the maintenance of cellular homoeostasis. In many cells, MT networks are comprised of centrosomal MTs and noncentrosomal MTs. However, it is unknown whether noncentrosomal MTs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service