Skip to Content
Merck
All Photos(4)

Documents

HPA020134

Sigma-Aldrich

Anti-URGCP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-AC004985.2-1, Anti-HBV X protein up-regulated gene 4 protein, Anti-HBxAg up-regulated gene 4 protein, Anti-Protein URG4, Anti-URG4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

LIPVLKIDHSHVLVKVSSTDSDSFVKRIRAIVGNVLRAPCRRVSVEDMAHAARKLGLKVDEDCEECQKAKDRMERITRKIKDSDAYRRDELRLQGDPWRKAAQVEKEFCQLQWAVDPPEKHRAE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... URGCP(55665)

General description

Upregulator of cell proliferation (URGCP) is a 104kDa protein and the gene associated with it is localized on human chromosome 7p13.

Immunogen

Protein URG4 recombinant protein epitope signature tag (PrEST)

Application

Anti-URGCP antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

Upregulator of cell proliferation (URGCP) is overexpressed in cervical cancer and it is associated with the growth of cancer cells. URGCP also enhances the growth of other carcinomas like hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74937

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guodong Yu et al.
OncoTargets and therapy, 9, 3059-3065 (2016-06-11)
URG4, a novel oncogene, is involved in the development and progression of various tumors. This study investigated the clinicopathological significance of URG4 in nasopharyngeal carcinoma (NPC). We used five NPC tissues and adjacent normal nasopharyngeal tissues to determine URG4 expression
Yesim Oymak et al.
Acta haematologica, 128(2), 73-76 (2012-06-09)
Elucidation of the molecular mechanisms of leukemogenesis is important for a better understanding of the prognosis of acute lymphoblastic leukemia (ALL). Studies have shown that the expression of upregulated gene 4 (URG4), which promotes cell growth and survival, is increased
Yavuz Dodurga et al.
Molecular biology reports, 39(12), 10173-10177 (2012-09-26)
Molecular targets in prostate cancer are continually being explored, for which there are currently few therapeutic options. Rapamycin (RPM) is an antifungal macrolide antibiotic isolated from Streptomyces hygroscopicus which can inhibit the G1 to S transition. URGCP (upregulator of cell
Lan Zhang et al.
BMC cancer, 14, 885-885 (2014-11-28)
Upregulator of cell proliferation 4 (URG4) has been implicated in the oncogenesis of certain cancers. However, the correlation between URG4 expression and clinicopathological significance in human cancer remains unclear. Therefore, this study investigated its expression and clinicopathological significance in cervical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service