Skip to Content
Merck
All Photos(2)

Documents

HPA020131

Sigma-Aldrich

Anti-PPIE antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cyclophilin E, Anti-Cyclophilin-33, Anti-PPIase E, Anti-Peptidyl-prolyl cis-trans isomerase E, Anti-Rotamase E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EEVDDKVLHAAFIPFGDITDIQIPLDYETEKHRGFAFVEFELAEDAAAAIDNMNESELFGRTIRVNLAKPMRIKEGSSRPVWSDDDWLKKF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPIE(10450)

General description

Peptidyl-prolyl cis-trans isomerase E (PPIE) is a cyclophilin which binds to RNA. It contains a conserved PPIase (peptidylprolyl isomerase) domain and a RNA-binding motif.

Immunogen

Peptidyl-prolyl cis-trans isomerase E recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Peptidyl-prolyl cis-trans isomerase E (PPIE) is involved in the cis-trans isomerization of the peptide bond before a proline residue. It facilitates conformational changes in folded and unfolded proteins. PPIE also mediates protein interactions and transport. PPIE recruits histone deacetylases to mixed lineage leukemia (MLL) genes and functions in down-regulating the target proteins like homeobox C8 (HOXC8). Studies have shown that it binds to influenza A virus nucleoprotein and prevents viral replication.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73809

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Wang et al.
FEBS letters, 582(5), 835-839 (2008-02-09)
Human nuclear cyclophilin 33 (hCyP33) was the first protein which was found to contain an RNA-binding motif and a PPIase domain. It was not known what cellular and physiological roles are played by the RNA-binding activity as well as the
Jing Chen et al.
Cancer research, 68(15), 6199-6207 (2008-08-05)
Reciprocal chromosomal translocations at the MLL gene locus result in expression of novel fusion proteins, such as MLL-ENL, associated with leukemia. The three PHD finger cassette, one of the highly conserved domains in MLL, is absent in all fusion proteins.
Sangho Park et al.
Biochemistry, 49(31), 6576-6586 (2010-08-04)
The mixed lineage leukemia (MLL) gene plays a critical role in epigenetic regulation of gene expression and is a frequent target of chromosomal translocations leading to leukemia. MLL plant homeodomain 3 (PHD3) is lost in all MLL translocation products, and
Robert A Hom et al.
Journal of molecular biology, 400(2), 145-154 (2010-05-13)
The nuclear protein cyclophilin 33 (Cyp33) is a peptidyl-prolyl cis-trans isomerase that catalyzes cis-trans isomerization of the peptide bond preceding a proline and promotes folding and conformational changes in folded and unfolded proteins. The N-terminal RNA-recognition motif (RRM) domain of
Zengfu Wang et al.
PloS one, 6(8), e22625-e22625 (2011-09-03)
The nucleoprotein (NP) of influenza A virus is a multifunctional protein that plays a critical role in the replication and transcription of the viral genome. Therefore, examining host factors that interact with NP may shed light on the mechanism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service