Skip to Content
Merck
All Photos(2)

Key Documents

HPA019055

Sigma-Aldrich

Anti-C21orf59 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-C21orf48, Anti-CILD26, Anti-FBB18, Anti-FLJ20467

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 481.00

CHF 481.00


Estimated to ship on17 May 2025



Select a Size

Change View
100 μL
CHF 481.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

CHF 481.00


Estimated to ship on17 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:1000- 1:2500

immunogen sequence

GDESQFLLQAPGSTELEELTVQVARVYNGRLKVQRLCSEMEELAEHGIFLPPNMQGLTDDQIEELKLKDEWGEKCVPSGGAVFKKDDIGRRNG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

In Chlamydomonas reinhardtii C21orf59 is localized exclusively to the flagellar matrix. It is cytoplasmic in rat trachea ciliated epithelial cells and colocalizes with spindle assembly abnormal protein-6 (SAS6), a centrosomal protein.

Immunogen

Uncharacterized protein C21orf59 recombinant protein epitope signature tag (PrEST)

Application

Anti-C21orf59 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

C21orf59 is crucial for cilia motility. Absence of C21orf59 is involved in primary ciliary dyskinesia (PCD). In affected humans cilia ultrastructure shows decrease in inner dynein arms, nexin links and microtubule disorganization. PCD patients have recurrent bronchitis, sinusitis and/or otitis media.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73869

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Austin-Tse et al.
American journal of human genetics, 93(4), 672-686 (2013-10-08)
Primary ciliary dyskinesia (PCD) is caused when defects of motile cilia lead to chronic airway infections, male infertility, and situs abnormalities. Multiple causative PCD mutations account for only 65% of cases, suggesting that many genes essential for cilia function remain to
Mathias Uhlén et al.
Molecular & cellular proteomics : MCP, 11(3), M111-M111 (2011-11-02)
The Human Proteome Project has been proposed to create a knowledge-based resource based on a systematical mapping of all human proteins, chromosome by chromosome, in a gene-centric manner. With this background, we here describe the systematic analysis of chromosome 21

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service