HPA011878
Anti-Ksr1 Antibody Produced In Rabbit
Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
KSR, RSU2
About This Item
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
product line
Prestige Antibodies® Powered by Atlas Antibodies
form
buffered aqueous glycerol solution
species reactivity
human
concentration
0.05mg/ml
technique(s)
immunofluorescence: 0.25-2 µg/ml
isotype
IgG
sequence note
SMIFGVKCKHCRLKCHNKCTKEAPACRISFLPLTRLRRTESVPSDINNPVDRAAEPHFGTLPKALTKKEHPPAMNHLDSSSNPSSTTSSTPSSPAPFPTSSNPSSATTPPNPSPGQRDSRFNFPA
Ensembl | human accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
-10 to -25°C
Gene Information
human ... KSR1(8844)
Immunogen
Application
The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies™ protocols and other useful information.
Features and Benefits
Physical form
Other Notes
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service