Skip to Content
Merck
All Photos(4)

Key Documents

HPA010729

Sigma-Aldrich

Anti-ABHD3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Abhydrolase domain-containing protein 3, Anti-Lung alpha/beta hydrolase 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 556.00

CHF 556.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μL
CHF 556.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

CHF 556.00


Estimated to ship on27 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KKPQLVTGGESFSRFLQDHCPVVTETYYPTVWCWEGRGQTLLRPFITSKPPVQYRNELIKTADGGQISLDWFDNDNSTCYMDASTRPTILLLPGLTGTSKESYILHMIHLSEELGYRCVVFNNRGVAGENLLTPRTYCCANTEDLE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABHD3(171586)

General description

ABHD3 (abhydrolase domain containing 3) belongs to the uncharacterized protein family called UPF0017. This protein contains α/β hydrolase fold, which is a catalytic domain found in multiple proteins. This gene is located at chromosome 18q11.1 and spans 54kb. ABHD3 shares high sequence similarity to ABHD1, and contains a single transmembrane domain at its N-terminal. This protein is expressed in the lung tissue.

Immunogen

Abhydrolase domain-containing protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The function of ABHD3 (abhydrolase domain containing 3) is yet to be characterized. However, studies suggest that this protein might function as a lipase, with specificity for medium-chain and oxidatively truncated phospholipids.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72088

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alasdair J Edgar et al.
Biochemical and biophysical research communications, 292(3), 617-625 (2002-04-02)
We have cloned 3 novel murine cDNAs encoding proteins containing an alpha/beta hydrolase fold; a catalytic domain found in a very wide range of enzymes. These proteins belong to the prosite UPF0017 uncharacterized protein family and we have named them
Jonathan Z Long et al.
Nature chemical biology, 7(11), 763-765 (2011-09-20)
All organisms, including humans, possess a huge number of uncharacterized enzymes. Here we describe a general cell-based screen for enzyme substrate discovery by untargeted metabolomics and its application to identify the protein α/β-hydrolase domain-containing 3 (ABHD3) as a lipase that
Alasdair J Edgar
BMC genomics, 4(1), 18-18 (2003-05-09)
Overlapping sense/antisense genes orientated in a tail-to-tail manner, often involving only the 3'UTRs, form the majority of gene pairs in mammalian genomes and can lead to the formation of double-stranded RNA that triggers the destruction of homologous mRNAs. Overlapping polyadenylation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service