Skip to Content
Merck
All Photos(7)

Key Documents

HPA008965

Sigma-Aldrich

Anti-GYPC antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD236 antigen antibody produced in rabbit, Anti-GLPC antibody produced in rabbit, Anti-GPD antibody produced in rabbit, Anti-Glycoconnectin antibody produced in rabbit, Anti-Glycophorin C antibody produced in rabbit, Anti-Glycophorin D antibody produced in rabbit, Anti-Glycoprotein β antibody produced in rabbit, Anti-PAS-2′ antibody produced in rabbit, Anti-Sialoglycoprotein D antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 556.00

CHF 556.00


Check Cart for Availability


Select a Size

Change View
100 μL
CHF 556.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

CHF 556.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RYMYRHKGTYHTNEAKGTEFAESADAALQGDPALQDAGDSSRKEYFI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GYPC(2995)

General description

GYPC gene contains four exons and codes for two sialoglycoproteins in humans, called glycophorin C and glycophorin D. This is an outcome of the presence of two start codons on the same transcript where translation is initiated. Glycophorin C (GYPC) protein is present on the surface of erythrocytes.

Immunogen

Glycophorin C recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Glycophorin C (GYPC) plays an essential role in the invasion of red blood cells (RBCs) by the malarial-causing parasite Plasmodium falciparum. It also functions as the receptor for PfEBP-2 (baebl, EBA-140), which in turn is the RBC-binding ligand of Plasmodium falciparum. This protein plays a key role in maintaining the mechanical stability of membrane and the shape of RBCs.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71967

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason A Wilder et al.
Molecular biology and evolution, 26(12), 2679-2687 (2009-08-15)
GYPC encodes two erythrocyte surface sialoglycoproteins in humans, glycophorin C and glycophorin D (GPC and GPD), via initiation of translation at two start codons on a single transcript. The malaria-causing parasite Plasmodium falciparum uses GPC as a means of invasion
Cheryl-Ann Lobo et al.
Blood, 101(11), 4628-4631 (2003-02-11)
We report in this paper that glycophorin C (GPC) is the receptor for PfEBP-2 (baebl, EBA-140), the newly identified erythrocyte binding ligand of Plasmodium falciparum. PfEBP-2 is a member of the Duffy binding-like erythrocyte binding protein (DBL-EBP) family. Although several
R Winardi et al.
Blood, 81(10), 2799-2803 (1993-05-15)
Human erythrocyte glycophorin C plays a functionally important role in maintaining erythrocyte shape and regulating membrane mechanical stability. We report here the characterization of the glycophorins C and D deficiency in erythrocytes of the Leach phenotype. Glycophorin C gene is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service