Skip to Content
Merck
All Photos(5)

Documents

HPA008751

Sigma-Aldrich

Anti-SMARCA5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SWI/SNF-related matrix- associated actin-dependent regulator of chromatin A5, Anti-SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 5, Anti-Sucrose nonfermenting protein 2 homolog, Anti-hSNF2H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SSAAEPPPPPPPESAPSKPAASIASGGSNSSNKGGPEGVAAQAVASAASAGPADAEMEEIFDDASPGKQKEIQEPDPTYEEKMQTDRANRFEYLLKQTELFAHFIQPA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMARCA5(8467)

General description

SMARCA5 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5) is a mammalian homolog of ISW1, and forms the predominant catalytic ATPase subunit of SWI-type chromatin remodeling complexes. This gene is localized to human chromosome 4q31.1→q31.2, and encodes a protein composed of 1052 amino acids.

Immunogen

SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 5 recombinant protein epitope signature tag (PrEST)

Application

Anti-SMARCA5 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

SMARCA5 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5) regulates the repair efficacy of transcription-hindering UV (ultraviolet) lesion, by promoting the interaction of CSB (Cockayne syndrome group B) to active TC-NER (transcription-coupled repair) complexes. This protein is associated with RNF168-dependent DNA damage signaling via DNA damage-mediated poly(ADP-ribosyl)ation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70943

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Özge Z Aydin et al.
Nucleic acids research, 42(13), 8473-8485 (2014-07-06)
Chromatin compaction of deoxyribonucleic acid (DNA) presents a major challenge to the detection and removal of DNA damage. Helix-distorting DNA lesions that block transcription are specifically repaired by transcription-coupled nucleotide excision repair, which is initiated by binding of the CSB
T Aihara et al.
Cytogenetics and cell genetics, 81(3-4), 191-193 (1998-09-08)
We have isolated a novel cDNA encoding a peptide with 86% sequence homology to hSNF2L protein, a previously isolated human homologue of Drosophila ISWI. This gene, designated SMARCA5, contained an open reading frame of 3,156 nucleotides encoding a 1,052 amino-acid
Godelieve Smeenk et al.
Journal of cell science, 126(Pt 4), 889-903 (2012-12-25)
Ionizing radiation (IR)-induced DNA double-strand breaks (DSBs) arising in native chromatin elicit an RNF8/RNF168-dependent ubiquitylation response, which triggers the recruitment of various repair factors. Precisely how this response is regulated in the context of chromatin remains largely unexplored. Here, we
Oxana Polyakova et al.
PloS one, 7(12), e51555-e51555 (2012-12-15)
SIRT6 is a member of the Sirtuin family of histone deacetylases that has been implicated in inflammatory, aging and metabolic pathways. Some of its actions have been suggested to be via physical interaction with NFκB and HIF1α and transcriptional regulation
Friederike L Pennemann et al.
Nature communications, 12(1), 7009-7009 (2021-12-03)
The cell intrinsic antiviral response of multicellular organisms developed over millions of years and critically relies on the ability to sense and eliminate viral nucleic acids. Here we use an affinity proteomics approach in evolutionary distant species (human, mouse and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service