Skip to Content
Merck
All Photos(4)

Documents

HPA007644

Sigma-Aldrich

Anti-HNRNPK antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HNRPK antibody produced in rabbit, Anti-Heterogeneous nuclear ribonucleoprotein K antibody produced in rabbit, Anti-TUNP antibody produced in rabbit, Anti-Transformation up-regulated nuclear protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

TEQPEETFPNTETNGEFGKRPAEDMEEEQAFKRSRNTDEMVELRILLQSKNAGAVIGKGGKNIKALRTDYNASVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNRNPK(3190)

General description

HNRNPK (heterogeneous nuclear ribonucleoprotein K) gene encodes a member of the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins that are associated with pre-mRNAs and mediate mRNA processing, metabolism and transport. The encoded protein has three repeats of KH domains that bind to RNA. It tenaciously binds to poly(C).

Immunogen

Heterogeneous nuclear ribonucleoprotein K recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HNRNPK (heterogeneous nuclear ribonucleoprotein K) protein is a nucleocytoplasmic shuttling protein that functions as a cofactor for p53 in mediating transcriptional induction of factors that regulate DNA repair, cell-cycle progression, or cell survival in response to DNA damage. Upon DNA damage, hnRNP K undergoes sumoylation through Pc2 E3 (E3 ligase) activity and activates p53 transcription. Deletion of this gene fails to induce transcription of p53 target genes and causes defects in DNA-damage-induced cell-cycle-checkpoint arrests.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71605

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Pelisch et al.
The Journal of biological chemistry, 287(36), 30789-30799 (2012-07-25)
Heterogeneous nuclear ribonucleoprotein (hnRNP) K is a nucleocytoplasmic shuttling protein that is a key player in the p53-triggered DNA damage response, acting as a cofactor for p53 in response to DNA damage. hnRNP K is a substrate of the ubiquitin
Abdeladim Moumen et al.
Cell, 123(6), 1065-1078 (2005-12-20)
In response to DNA damage, mammalian cells trigger the p53-dependent transcriptional induction of factors that regulate DNA repair, cell-cycle progression, or cell survival. Through differential proteomics, we identify heterogeneous nuclear ribonucleoprotein K (hnRNP K) as being rapidly induced by DNA
Ravindra Varma Polisetty et al.
Scientific reports, 6, 26882-26882 (2016-06-02)
Diffuse astrocytoma (DA; WHO grade II) is a low-grade, primary brain neoplasm with high potential of recurrence as higher grade malignant form. We have analyzed differentially expressed membrane proteins from these tumors, using high-resolution mass spectrometry. A total of 2803

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service