Skip to Content
Merck
All Photos(1)

Key Documents

HPA003195

Sigma-Aldrich

Anti-CILP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CILP-1 antibody produced in rabbit, Anti-Cartilage intermediate layer protein 1 precursor antibody produced in rabbit, Anti-Cartilage intermediate-layer protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

YKHESKLVLRKLQQHQAGEYFCKAQSDAGAVKSKVAQLIVTASDETPCNPVPESYLIRLPHDCFQNATNSFYYDVGRCPVKTCAGQQDNGIRCRDAVQNCCGISKTEEREIQCSGYTLPTK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CILP(8483)

General description

The CILP (Cartilage intermediate layer protein 1) gene encodes a 91.5kDa protein that is expressed abundantly in intervertebral discs and its levels are increased in early osteoarthrosis cartilage. The CILP gene is mapped to human chromosome 15q22.

Immunogen

Cartilage intermediate layer protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CILP (Cartilage intermediate layer protein 1) gene encodes a protein that may function in cartilage repair and degradation. It functions as an antagonist to IGF-1 (insulin-like growth factor 1) and inhibits IGF1-induced proliferation and sulfated proteoglycan synthesis. Overexpression of this protein leads to inorganic pyrophosphate (PPi) supersaturation in aging and osteoarthritis cartilage. It inhibits TGF-β1 signaling and TGF-β1-mediated induction of cartilage matrix genes by interacting with TGF-β1. This has an important role in etiology and pathogenesis of lumbar disc disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79908

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoji Seki et al.
Nature genetics, 37(6), 607-612 (2005-05-03)
Lumbar disc disease (LDD) is caused by degeneration of intervertebral discs of the lumbar spine. One of the most common musculoskeletal disorders, LDD has strong genetic determinants. Using a case-control association study, we identified a functional SNP (1184T --> C
P Lorenzo et al.
The Journal of biological chemistry, 273(36), 23463-23468 (1998-08-29)
A novel, somewhat basic noncollagenous protein was purified from guanidine hydrochloride extracts of human articular cartilage using cesium chloride density gradient centrifugation, followed by ion-exchange chromatography at pH 5, and gel filtration on two serially coupled columns of Superose 6
Kristen Johnson et al.
Arthritis and rheumatism, 48(5), 1302-1314 (2003-05-15)
Aging and osteoarthritic (OA) cartilage commonly demonstrate enhanced expression of the large, transforming growth factor beta (TGFbeta)-inducible glycoprotein cartilage intermediate-layer protein (CILP) as well as enhanced extracellular inorganic pyrophosphate (PPi) that promotes the deposition of calcium pyrophosphate dihydrate crystals. In
P Lorenzo et al.
Matrix biology : journal of the International Society for Matrix Biology, 18(5), 445-454 (1999-12-22)
The human cDNA for cartilage intermediate layer protein (CILP) codes for a larger precursor protein that consists of CILP and a homologue to porcine Nucleotide pyrophosphohydrolase (NTPPHase) [Lorenzo et al. 1998a. J. Biol. Chem. 273, 23469-23475]. The human gene has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service