Skip to Content
Merck
All Photos(1)

Key Documents

HPA003124

Sigma-Aldrich

Anti-FANCB antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FAAP95 antibody produced in rabbit, Anti-Fanconi anemia group B protein antibody produced in rabbit, Anti-Fanconi anemia-associated polypeptide of 95 kDa antibody produced in rabbit, Anti-Protein FACB antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

DSLNSDCLTSFKITDLGKINYSSEPSDCNEDDLFEDKQENRYLVVPPLETGLKVCFSSFRELRQHLLLKEKIISKSYKALINLVQGKDDNTSSAEEKECLVPLCGEEENSVHILDEKLSDNFQDSEQLVEKIWY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FANCB(2187)

General description

FANCB (Fanconi anemia, complementation group B) is a part of the five protein containing Fanconi anemia core complex, which also contains four unidentified parts. It has a molecular weight of 95kDa. This gene is localized to human chromosome Xp22.31.

Immunogen

Fanconi anemia group B protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FANCB (Fanconi anemia, complementation group B) forms an essential component of the nuclear protein ′core complex′ that functions in monoubiquitination of FANCD2, which is a crucial step in the DNA-damage response pathway linked to Fanconi anemia and BRCA. Mutations in this gene cause Fanconi anemia that is characterized by anemia, leukopenia and thrombopenia. It also results in hypersensitivity to DNA-damaging agents, chromosomal instability and defective DNA repair. Severe cases cause X linked VACTERL-H syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74432

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S T Holden et al.
Journal of medical genetics, 43(9), 750-754 (2006-05-09)
The VACTERL with hydrocephalus (VACTERL-H) phenotype is recognised to be a severe manifestation of autosomal recessive Fanconi anaemia. Several families have been described in which the VACTERL-H phenotype segregates as an X linked syndrome. The mutations which cause X linked
Amom Ruhikanta Meetei et al.
Nature genetics, 36(11), 1219-1224 (2004-10-27)
Fanconi anemia is an autosomal recessive syndrome characterized by diverse clinical symptoms, hypersensitivity to DNA crosslinking agents, chromosomal instability and susceptibility to cancer. Fanconi anemia has at least 11 complementation groups (A, B, C, D1, D2, E, F, G, I

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service