Skip to Content
Merck
All Photos(5)

Key Documents

HPA000903

Sigma-Aldrich

Anti-AHSA1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AHA1 antibody produced in rabbit, Anti-Activator of 90 kDa heat shock protein ATPase homolog 1 antibody produced in rabbit, Anti-p38 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 481.00

CHF 481.00


Estimated to ship on16 May 2025



Select a Size

Change View
100 μL
CHF 481.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

CHF 481.00


Estimated to ship on16 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

PTMNGESVDPVGQPALKTEERKAKPAPSKTQARPVGVKIPTCKITLKETFLTSPEELYRVFTTQELVQAFTHAPATLEADRGGKFHMVDGNVSGEFTDLVPEKHIVMKWRFKSWPEGHFATITLTFIDKNGETELCMEGRGIPAPEEE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AHSA1(10598)

Immunogen

Activator of 90 kDa heat shock protein ATPase homolog 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Activator of 90kDa heat shock protein ATPase homolog 1 is an enzyme encoded by the AHSA1 gene in humans. Overexpression of this protein in endothelial cells (ECs) promotes association of endothelial NO synthase (eNOS) and heat-shock protein 90 (Hsp90), phosphorylation of Ser-1179 of eNOS, increases NO production and cell migration. It regulates VEGF signaling to eNOS and angiogenesis through modulation of Hsp90 activity. Aha1 may play important role in modulating RNA splicing and DNA repair along with other cellular processes. Its activity integrates chaperone function with client folding energetics by modulating ATPase sensitive N-terminal dimer structural transitions and protects transient folding intermediates. It may also contribute to protein misfolding systems disorders. Aha1 increased levels are associated with enhanced interactions with Hsp90 that results in increased Hsp90 ATPase activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70424

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atanas V Koulov et al.
Molecular biology of the cell, 21(6), 871-884 (2010-01-22)
The activator of Hsp90 ATPase 1, Aha1, has been shown to participate in the Hsp90 chaperone cycle by stimulating the low intrinsic ATPase activity of Hsp90. To elucidate the structural basis for ATPase stimulation of human Hsp90 by human Aha1
Sachiyo Okayama et al.
The Journal of biological chemistry, 289(10), 6513-6525 (2014-01-24)
The p53 tumor suppressor gene encodes a homotetrameric transcription factor which is activated in response to a variety of cellular stressors, including DNA damage and oncogene activation. p53 mutations occur in >50% of human cancers. Although p53 has been shown
Liang Sun et al.
Biochimica et biophysica acta, 1823(6), 1092-1101 (2012-04-17)
The activator of Hsp90 ATPase, Aha1, is an Hsp90 co-chaperone that has been suggested to act as a general stimulator of Hsp90 function. In this report, we have characterized the interaction of Aha1 with Hsp90 and its co-chaperones in rabbit
Fanny Desjardins et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(10), 2484-2492 (2012-08-04)
Vascular endothelial growth factor (VEGF) signaling to endothelial NO synthase (eNOS) plays a central role in angiogenesis. In endothelial cells (ECs), heat-shock protein 90 (Hsp90) is also a regulator of eNOS activity. Our study is designed to determine whether modulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service