Skip to Content
Merck
All Photos(2)

Key Documents

HISTO-1

Sigma-Aldrich

Visikol® HISTO-1

Sign Into View Organizational & Contract Pricing

Select a Size

30 ML
CHF 335.00
100 ML
CHF 714.00

CHF 335.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
30 ML
CHF 335.00
100 ML
CHF 714.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.47

CHF 335.00


Check Cart for Availability

Request a Bulk Order

shipped in

ambient

storage temp.

room temp

General description

Visikol HISTO-1 is an optical clearing agent for biological tissues enabling rapid and easy visualization of tissues in 3D without altering the underlying cellular morphology. Visikol HISTO-1 is fully compatible with immunostaining as well as many common other common fluorescent labels. Using Visikol HISTO-1 with confocal or light sheet microscopy allows researchers to see deeper into tissues than ever before.

Application

  • Optical clearing agent in tissue imaging studies
  • Refractive index matching agent for microscopy

Visikol HISTO products are easy-to-use and ideal for rapid tissue clearing applications. Tissue clearing is completely reversible with no alteration to the tissue.

Features and Benefits

  • Render tissue samples transparent for imaging.
  • Achieve clearer, sharper images in antibody tissue staining applications.
  • The Visikol HISTO tissue clearing process does not affect prior immunolabeling.
  • Save time. The Visikol HISTO method is rapid relative to many popular current tissue clearing techniques.
  • Retain morphology and lipid content in your samples.
  • The HISTO tissue clearing method is easily reversible for follow up 2D histology/IHC or for 3D data validation against traditional histology.
  • Non-toxic.

Legal Information

HISTO-1 is a trademark of Visikol, Inc.
Visikol is a registered trademark of Visikol, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Visikol® HISTOTM tissue clearing solutions enable developmental biology, neuroscience, and cancer research with reversible protocols.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service