Skip to Content
Merck
All Photos(1)

Key Documents

H5789

Sigma-Aldrich

Anti-HuD antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ELAVL4, Anti-Human Antigen D

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
CHF 584.00

CHF 584.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
200 μL
CHF 584.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 584.00


Estimated to ship on26 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40 kDa

species reactivity

human, mouse

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10-20 μg/mL using heat retrieved, human intestine tissue
western blot (chemiluminescent): 5-10 μg/mL using mouse brain extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELAVL4(1996)
mouse ... Elavl4(15572)

Related Categories

General description

Hu antigen D (HuD) protein is highly conserved and a neuron-specific RNA-binding protein. This protein belongs to the embryonic lethal, abnormal vision, Drosophila-like (ELAVL)/Hu family. The HuD gene is located on the human chromosome at 1p33-p32.3.

Specificity

Anti-HuD specifically recognizes human and mouse HuD.

Immunogen

synthetic peptide corresponding to amino acids 240-255 of human HuD with an N-terminal added cysteine, conjugated to KLH.

Application

Anti-HuD antibody produced in rabbit may be used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Hu antigen D (HuD) protein binds to the 3′ untranslated region (UTR) of mRNA of genes like N-myc gene and growth-associated protein (GAP43). This binding results in an increase in the half-life of these mRNA. HuD plays a vital role in several regulatory processes, such as the processing of pre-mRNA, mRNA stability and translation. This protein is also involved in neuronal functioning and development. HuD protein also plays a role in neuronal plasticity, which includes, learning and memory, recovery from axonal injury and multiple neurological diseases.

Target description

HuD protein binds to the 3′ UTR of mRNA of geneslike N-myc gene and GAP43, and increases the half-life ofthese mRNA.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide and 1% inactivated bovine serum albumin.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Chromosomal Aberrations in the Head and Neck Squamous Cell Carcinoma Cell Lines: PCI-1, PCI-13 and PCI-50
Moon Y E, et al.
Korean Journal of Otorhinolaryngology - Head and Neck Surgery, 52(9), 741-750 (2009)
Lucas M Bronicki et al.
RNA (New York, N.Y.), 19(8), 1019-1037 (2013-07-19)
Precise control of messenger RNA (mRNA) processing and abundance are increasingly being recognized as critical for proper spatiotemporal gene expression, particularly in neurons. These regulatory events are governed by a large number of trans-acting factors found in neurons, most notably
Feifei Wang et al.
Anticancer research, 37(8), 4515-4522 (2017-07-26)
The ELAVL4 (HuD) is a neuron-specific RNA-binding protein expressed in 100% of small cell lung cancer (SCLC) cells and over 50% of neuroblastoma cells. The aim of this study was to investigate the serum HuD concentration in SCLC patients and
Riccardo De Santis et al.
Cell reports, 27(13), 3818-3831 (2019-06-27)
Amyotrophic lateral sclerosis (ALS) has been genetically linked to mutations in RNA-binding proteins (RBPs), including FUS. Here, we report the RNA interactome of wild-type and mutant FUS in human motor neurons (MNs). This analysis identified a number of RNA targets.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service