Skip to Content
Merck
All Photos(1)

Key Documents

H4395

Sigma-Aldrich

N-Heptyl-5-Chloro-1-Naphthalenesulfonamide

powder

Synonym(s):

Protein kinase C activator, SC-10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22ClNO2S
CAS Number:
Molecular Weight:
339.88
MDL number:
UNSPSC Code:
12171500

form

powder

color

white

solubility

DMSO: 10 mM
ethanol: soluble

storage temp.

−20°C

SMILES string

CCCCCCCNS(=O)(=O)c1cccc2c(Cl)cccc12


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ito et al.
Biochemistry, 25(15), 4179-4184 (1986-07-29)
Naphthalenesulfonamide derivatives were used to study the mechanism of regulation of Ca2+-dependent smooth muscle myosin light chain phosphorylation catalyzed by Ca2+-activated, phospholipid-dependent protein kinase (protein kinase C) and myosin light chain kinase. Derivatives such as N-(6-phenylhexyl)-5-chloro-1-naphthalenesulfonamide (SC-9), with a hydrophobic
H Nishino et al.
Biochimica et biophysica acta, 889(2), 236-239 (1986-11-28)
N-(6-Phenylhexyl)-5-chloro-1-naphthalenesulfonamide (SC-9) activated Ca2+-activated, phospholipid-dependent protein kinase (protein kinase C). SC-9 acted as a substitute for phosphatidylserine, which is one of the endogenous factors in activating protein kinase C. SC-9 was also effective in regulating the physiological functions at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service