Skip to Content
Merck
All Photos(1)

Key Documents

H3292

Sigma-Aldrich

Histo/Zyme, Ready to Use, Antigen Retriever

Sign Into View Organizational & Contract Pricing

Select a Size

15 ML
CHF 261.00
50 ML
CHF 704.00

CHF 261.00


Estimated to ship on03 May 2025


Request a Bulk Order

Select a Size

Change View
15 ML
CHF 261.00
50 ML
CHF 704.00

About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

CHF 261.00


Estimated to ship on03 May 2025


Request a Bulk Order

application(s)

hematology
histology

Quality Level

storage temp.

2-8°C

Application

Histo/Zyme is a stabilized proteolytic enzyme in buffered solution, used to unmask immunoreactive sites altered by fixation and/or the embedding process. Treatment with a proteolytic enzyme in immunohistochemistry (IHC) is required to digest excess linkages and expose the antigen. The use of Histo/Zyme results in superior staining over traditionally used enzyme pretreatments, such as pepsin and trypsin. Moreover, Histo/Zyme requires only a brief incubation at room temperature. Performing antigen recovery with Histo/Zyme will allow you to dilute the primary antibody 5 to 10 fold further and achieve optimal staining.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service